Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 163920 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HMGN1 - high mobility group nucleosome binding domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004965.6 | Intron | NP_004956.5 | ||||
XM_011529543.1 | Intron | XP_011527845.1 | ||||
XM_011529544.1 | Intron | XP_011527846.1 | ||||
XM_011529545.1 | Intron | XP_011527847.1 | ||||
XM_011529547.1 | Intron | XP_011527849.1 | ||||
XM_011529549.1 | Intron | XP_011527851.1 | ||||
XM_017028332.1 | Intron | XP_016883821.1 | ||||
XM_017028333.1 | Intron | XP_016883822.1 | ||||
XM_017028334.1 | Intron | XP_016883823.1 | ||||
XM_017028335.1 | Intron | XP_016883824.1 | ||||
XM_017028336.1 | Intron | XP_016883825.1 | ||||
XM_017028338.1 | Intron | XP_016883827.1 |
Set Membership: |
HapMap |