Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603385 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NAE1 - NEDD8 activating enzyme E1 subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018159.1 | Intron | NP_001018169.1 | ||||
NM_001018160.1 | Intron | NP_001018170.1 | ||||
NM_001286500.1 | Intron | NP_001273429.1 | ||||
NM_003905.3 | Intron | NP_003896.1 | ||||
XM_005256215.1 | Intron | XP_005256272.1 | ||||
XM_011523422.2 | Intron | XP_011521724.1 |
TERB1 - telomere repeat binding bouquet formation protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |