Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607576 MIM: 609303 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CECR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CECR2 - CECR2, histone acetyl-lysine reader | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A18 - solute carrier family 25 member 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303484.1 | Intron | NP_001290413.1 | ||||
NM_031481.2 | Intron | NP_113669.1 | ||||
XM_011546149.2 | Intron | XP_011544451.1 | ||||
XM_011546150.2 | Intron | XP_011544452.1 | ||||
XM_011546151.2 | Intron | XP_011544453.1 | ||||
XM_011546152.1 | Intron | XP_011544454.1 | ||||
XM_011546153.2 | Intron | XP_011544455.1 | ||||
XM_011546154.2 | Intron | XP_011544456.1 | ||||
XM_017028968.1 | Intron | XP_016884457.1 | ||||
XM_017028969.1 | Intron | XP_016884458.1 | ||||
XM_017028970.1 | Intron | XP_016884459.1 | ||||
XM_017028971.1 | Intron | XP_016884460.1 | ||||
XM_017028972.1 | Intron | XP_016884461.1 | ||||
XM_017028973.1 | Intron | XP_016884462.1 | ||||
XM_017028974.1 | Intron | XP_016884463.1 | ||||
XM_017028975.1 | Intron | XP_016884464.1 |
Set Membership: |
HapMap |