Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609360 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNLS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNLS - renalase, FAD dependent amine oxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031709.2 | Intron | NP_001026879.2 | ||||
NM_018363.3 | Intron | NP_060833.1 | ||||
XM_005269946.1 | Intron | XP_005270003.1 | ||||
XM_005269947.1 | Intron | XP_005270004.1 | ||||
XM_005269948.2 | Intron | XP_005270005.1 | ||||
XM_005269949.4 | Intron | XP_005270006.1 | ||||
XM_005269950.3 | Intron | XP_005270007.1 | ||||
XM_011539924.2 | Intron | XP_011538226.1 | ||||
XM_011539927.2 | Intron | XP_011538229.1 | ||||
XM_017016380.1 | Intron | XP_016871869.1 | ||||
XM_017016381.1 | Intron | XP_016871870.1 | ||||
XM_017016382.1 | Intron | XP_016871871.1 | ||||
XM_017016383.1 | Intron | XP_016871872.1 | ||||
XM_017016384.1 | Intron | XP_016871873.1 | ||||
XM_017016385.1 | Intron | XP_016871874.1 |
Set Membership: |
HapMap |