Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
84 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604704 MIM: 608848 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAR3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCAR3 - breast cancer anti-estrogen resistance 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261408.1 | Intron | NP_001248337.1 | ||||
NM_001261409.1 | Intron | NP_001248338.1 | ||||
NM_001261410.1 | Intron | NP_001248339.1 | ||||
NM_001308251.1 | Intron | NP_001295180.1 | ||||
NM_003567.3 | Intron | NP_003558.1 | ||||
XM_011542251.2 | Intron | XP_011540553.1 | ||||
XM_017002480.1 | Intron | XP_016857969.1 |
FNBP1L - formin binding protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024948.2 | Intron | NP_001020119.1 | ||||
NM_001164473.2 | Intron | NP_001157945.1 | ||||
NM_017737.4 | Intron | NP_060207.2 | ||||
XM_011541625.2 | Intron | XP_011539927.1 | ||||
XM_017001533.1 | Intron | XP_016857022.1 |