Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603690 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC33A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC33A1 - solute carrier family 33 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190992.1 | Intron | NP_001177921.1 | ||||
NM_004733.3 | Intron | NP_004724.1 | ||||
XM_011513311.2 | Intron | XP_011511613.1 | ||||
XM_017007462.1 | Intron | XP_016862951.1 | ||||
XM_017007463.1 | Intron | XP_016862952.1 | ||||
XM_017007464.1 | Intron | XP_016862953.1 |