Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608039 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFESD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RFESD - Rieske Fe-S domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131065.1 | 3411 | Intron | NP_001124537.1 | |||
NM_001131066.1 | 3411 | Intron | NP_001124538.1 | |||
NM_173362.3 | 3411 | Intron | NP_775498.1 | |||
XM_011543360.2 | 3411 | UTR 3 | XP_011541662.1 | |||
XM_011543361.2 | 3411 | UTR 3 | XP_011541663.1 | |||
XM_017009413.1 | 3411 | UTR 3 | XP_016864902.1 |
SPATA9 - spermatogenesis associated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031952.3 | 3411 | Intron | NP_114158.2 | |||
XM_011543664.1 | 3411 | Intron | XP_011541966.1 | |||
XM_011543666.1 | 3411 | Intron | XP_011541968.1 | |||
XM_011543667.2 | 3411 | Intron | XP_011541969.1 | |||
XM_017009948.1 | 3411 | Intron | XP_016865437.1 | |||
XM_017009949.1 | 3411 | Intron | XP_016865438.1 | |||
XM_017009950.1 | 3411 | Intron | XP_016865439.1 | |||
XM_017009951.1 | 3411 | Intron | XP_016865440.1 |
Set Membership: |
HapMap |