Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612962 MIM: 604034 MIM: 602098 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCTN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DCTN5 - dynactin subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ERN2 - endoplasmic reticulum to nucleus signaling 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLK1 - polo like kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005030.5 | Intron | NP_005021.2 |
Set Membership: |
HapMap Validated |