Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610507 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LEO1 - LEO1 homolog, Paf1/RNA polymerase II complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286430.1 | 2055 | UTR 3 | NP_001273359.1 | |||
NM_001323903.1 | 2055 | UTR 3 | NP_001310832.1 | |||
NM_001323904.1 | 2055 | UTR 3 | NP_001310833.1 | |||
NM_138792.3 | 2055 | UTR 3 | NP_620147.1 | |||
XM_017021911.1 | 2055 | UTR 3 | XP_016877400.1 |