Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WSCD2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WSCD2 - WSC domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304447.1 | Intron | NP_001291376.1 | ||||
NM_014653.3 | Intron | NP_055468.2 | ||||
XM_017020243.1 | Intron | XP_016875732.1 | ||||
XM_017020244.1 | Intron | XP_016875733.1 | ||||
XM_017020245.1 | Intron | XP_016875734.1 | ||||
XM_017020246.1 | Intron | XP_016875735.1 | ||||
XM_017020247.1 | Intron | XP_016875736.1 | ||||
XM_017020248.1 | Intron | XP_016875737.1 | ||||
XM_017020249.1 | Intron | XP_016875738.1 | ||||
XM_017020250.1 | Intron | XP_016875739.1 | ||||
XM_017020251.1 | Intron | XP_016875740.1 | ||||
XM_017020252.1 | Intron | XP_016875741.1 |