Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612662 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRS2P2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MRS2P2 - MRS2 pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D15 - TBC1 domain family member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146213.1 | Intron | NP_001139685.2 | ||||
NM_001146214.1 | Intron | NP_001139686.1 | ||||
NM_022771.4 | Intron | NP_073608.4 | ||||
XM_006719564.2 | Intron | XP_006719627.1 | ||||
XM_006719565.3 | Intron | XP_006719628.1 | ||||
XM_006719566.2 | Intron | XP_006719629.1 | ||||
XM_006719567.2 | Intron | XP_006719630.1 | ||||
XM_011538679.1 | Intron | XP_011536981.1 | ||||
XM_011538680.1 | Intron | XP_011536982.1 | ||||
XM_011538681.2 | Intron | XP_011536983.1 | ||||
XM_011538682.1 | Intron | XP_011536984.1 | ||||
XM_017019824.1 | Intron | XP_016875313.1 | ||||
XM_017019825.1 | Intron | XP_016875314.1 | ||||
XM_017019826.1 | Intron | XP_016875315.1 | ||||
XM_017019827.1 | Intron | XP_016875316.1 | ||||
XM_017019828.1 | Intron | XP_016875317.1 |
Set Membership: |
HapMap Validated |