Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616976 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM40 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TRIM40 - tripartite motif containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286633.1 | 787 | UTR 5 | NP_001273562.1 | |||
NM_138700.4 | 787 | UTR 5 | NP_619645.1 | |||
XM_011514305.1 | 787 | UTR 5 | XP_011512607.1 | |||
XM_011514306.1 | 787 | UTR 5 | XP_011512608.1 | |||
XM_011514308.1 | 787 | UTR 5 | XP_011512610.1 | |||
XM_011514309.1 | 787 | UTR 5 | XP_011512611.1 |
Set Membership: |
HapMap Validated |