Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607013 MIM: 608737 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BRF2 - BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB11FIP1 - RAB11 family interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002814.2 | 7200 | UTR 3 | NP_001002814.2 | |||
NM_025151.4 | 7200 | UTR 3 | NP_079427.4 | |||
XM_017013869.1 | 7200 | Intron | XP_016869358.1 |