Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGCACATAATCAGAGAAGTGA[C/G]GCGGCCATGCCGAGGTATACGTGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608958 MIM: 604932 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADA - adenosine deaminase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000022.3 | 1416 | UTR 3 | NP_000013.2 | |||
NM_001322050.1 | 1416 | UTR 3 | NP_001308979.1 | |||
NM_001322051.1 | 1416 | UTR 3 | NP_001308980.1 |
PKIG - protein kinase (cAMP-dependent, catalytic) inhibitor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281444.1 | 1416 | Intron | NP_001268373.1 | |||
NM_001281445.1 | 1416 | Intron | NP_001268374.1 | |||
NM_007066.4 | 1416 | Intron | NP_008997.1 | |||
NM_181804.2 | 1416 | Intron | NP_861520.1 | |||
NM_181805.2 | 1416 | Intron | NP_861521.1 | |||
XM_017027614.1 | 1416 | Intron | XP_016883103.1 | |||
XM_017027615.1 | 1416 | Intron | XP_016883104.1 | |||
XM_017027616.1 | 1416 | Intron | XP_016883105.1 |
Set Membership: |
HapMap |