Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTAATACTTTACTAATTTTGG[C/T]TATTTCAATTATCTACATTTGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613594 MIM: 613595 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTN3A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BTN3A3 - butyrophilin subfamily 3 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242803.1 | Intron | NP_001229732.1 | ||||
NM_006994.4 | Intron | NP_008925.1 | ||||
NM_197974.2 | Intron | NP_932078.2 |
Set Membership: |
HapMap |