Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACCCAGTGGCGGGCATCGCGGG[C/T]TCGGCGGCCAAGAGCGTGCGGCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133450 MIM: 602128 MIM: 602220 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EWSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EWSR1 - EWS RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GAS2L1 - growth arrest specific 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278730.1 | 208 | Silent Mutation | GGC,GGT | G,G 10 | NP_001265659.1 | |
NM_006478.4 | 208 | Silent Mutation | GGC,GGT | G,G 10 | NP_006469.2 | |
NM_152236.2 | 208 | Intron | NP_689422.1 | |||
XM_011529824.1 | 208 | Silent Mutation | GGC,GGT | G,G 10 | XP_011528126.1 | |
XM_011529825.1 | 208 | Silent Mutation | GGC,GGT | G,G 10 | XP_011528127.1 | |
XM_017028533.1 | 208 | Silent Mutation | GGC,GGT | G,G 10 | XP_016884022.1 |
RASL10A - RAS like family 10 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |