Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCTGAGCCCTGAGCCCCTGTTC[C/G]CAAGGAGCAGAGTCCTTCTCAGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM193A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM193A - family with sequence similarity 193 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256666.1 | Intron | NP_001243595.1 | ||||
NM_001256667.1 | Intron | NP_001243596.1 | ||||
NM_001256668.1 | Intron | NP_001243597.1 | ||||
NM_003704.3 | Intron | NP_003695.3 | ||||
XM_006713930.2 | Intron | XP_006713993.2 | ||||
XM_006713932.2 | Intron | XP_006713995.2 | ||||
XM_011513590.1 | Intron | XP_011511892.1 | ||||
XM_011513593.2 | Intron | XP_011511895.1 | ||||
XM_017008788.1 | Intron | XP_016864277.1 |