Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138981 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSF2RB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CSF2RB - colony stimulating factor 2 receptor beta common subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000395.2 | Intron | NP_000386.1 | ||||
XM_005261340.3 | Intron | XP_005261397.1 | ||||
XM_011529903.2 | Intron | XP_011528205.1 | ||||
XM_011529904.2 | Intron | XP_011528206.1 | ||||
XM_011529905.2 | Intron | XP_011528207.1 |
Set Membership: |
HapMap Validated |