Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAATTTGTAGCAAAATTACTAAA[A/C]GTGAACAGGCTGTTTGAGGTAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615902 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM177A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM177A1 - family with sequence similarity 177 member A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927178 - uncharacterized LOC101927178 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP2R3C - protein phosphatase 2 regulatory subunit B''gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305155.1 | Intron | NP_001292084.1 | ||||
NM_001305156.1 | Intron | NP_001292085.1 | ||||
NM_017917.3 | Intron | NP_060387.2 | ||||
XM_005267782.3 | Intron | XP_005267839.1 | ||||
XM_017021388.1 | Intron | XP_016876877.1 | ||||
XM_017021389.1 | Intron | XP_016876878.1 |
Set Membership: |
HapMap |