Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
67 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191510 MIM: 164790 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSDE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CSDE1 - cold shock domain containing E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007553.2 | 3714 | UTR 3 | NP_001007554.1 | |||
NM_001130523.2 | 3714 | UTR 3 | NP_001123995.1 | |||
NM_001242891.1 | 3714 | UTR 3 | NP_001229820.1 | |||
NM_001242892.1 | 3714 | UTR 3 | NP_001229821.1 | |||
NM_001242893.1 | 3714 | UTR 3 | NP_001229822.1 | |||
NM_007158.5 | 3714 | UTR 3 | NP_009089.4 |
NRAS - neuroblastoma RAS viral oncogene homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002524.4 | 3714 | Intron | NP_002515.1 |
Set Membership: |
HapMap |