Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGCCCTCACCCCAACATGCCA[C/G]AATCTCCTTCCTCTCCAGCTTGTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608740 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFAM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NFAM1 - NFAT activating protein with ITAM motif 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318323.1 | 2180 | UTR 3 | NP_001305252.1 | |||
NM_145912.6 | 2180 | UTR 3 | NP_666017.1 | |||
XM_005261374.4 | 2180 | UTR 3 | XP_005261431.1 |