Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTATGCTTACTTCCCTTAAAAT[A/G]TGAGGTTAACAATTTACATTGACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609132 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM1A - lysine demethylase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009999.2 | Intron | NP_001009999.1 | ||||
NM_015013.3 | Intron | NP_055828.2 | ||||
XM_005245786.2 | Intron | XP_005245843.1 | ||||
XM_006710472.3 | Intron | XP_006710535.1 | ||||
XM_006710473.2 | Intron | XP_006710536.1 | ||||
XM_006710474.3 | Intron | XP_006710537.1 | ||||
XM_017000716.1 | Intron | XP_016856205.1 | ||||
XM_017000717.1 | Intron | XP_016856206.1 | ||||
XM_017000718.1 | Intron | XP_016856207.1 | ||||
XM_017000719.1 | Intron | XP_016856208.1 | ||||
XM_017000720.1 | Intron | XP_016856209.1 |