Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAAGGGAGGGCTGAGCTGTGGG[A/C]TAAACTGCAGGCACCGGTCTACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608359 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373057 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC105373057 - DNA-directed RNA polymerase II subunit RPB1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT4A1 - sulfotransferase family 4A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014351.3 | Intron | NP_055166.1 | ||||
XM_011530120.2 | Intron | XP_011528422.1 | ||||
XM_011530121.1 | Intron | XP_011528423.1 |
Set Membership: |
HapMap Validated |