Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAATTTTCAGGAAACTGTTGAGT[C/T]AGTAGGTAGAATATTGAATGTTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929280 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929280 - uncharacterized LOC101929280 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB40C - RAB40C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172663.1 | Intron | NP_001166134.1 | ||||
NM_001172664.1 | Intron | NP_001166135.1 | ||||
NM_001172665.1 | Intron | NP_001166136.1 | ||||
NM_001172666.1 | Intron | NP_001166137.1 | ||||
NM_021168.4 | Intron | NP_066991.3 |
Set Membership: |
HapMap |