Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606750 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBP1 - Z-DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160417.1 | 1423 | Intron | NP_001153889.1 | |||
NM_001160418.1 | 1423 | Intron | NP_001153890.1 | |||
NM_001160419.2 | 1423 | Intron | NP_001153891.1 | |||
NM_001323966.1 | 1423 | Intron | NP_001310895.1 | |||
NM_030776.2 | 1423 | Intron | NP_110403.2 | |||
XM_011529057.2 | 1423 | UTR 3 | XP_011527359.1 | |||
XM_011529058.2 | 1423 | Intron | XP_011527360.1 | |||
XM_017028086.1 | 1423 | Intron | XP_016883575.1 |