Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PSMF1 - proteasome inhibitor subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM74B - transmembrane protein 74B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304748.1 | Intron | NP_001291677.1 | ||||
NM_001304749.1 | Intron | NP_001291678.1 | ||||
NM_018354.2 | Intron | NP_060824.1 | ||||
XM_011529281.2 | Intron | XP_011527583.1 | ||||
XM_011529282.2 | Intron | XP_011527584.1 | ||||
XM_011529283.2 | Intron | XP_011527585.1 | ||||
XM_011529284.2 | Intron | XP_011527586.1 | ||||
XM_011529285.2 | Intron | XP_011527587.1 | ||||
XM_017027923.1 | Intron | XP_016883412.1 | ||||
XM_017027924.1 | Intron | XP_016883413.1 | ||||
XM_017027925.1 | Intron | XP_016883414.1 |