Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613542 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTRF1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTRF1L - mitochondrial translational release factor 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114184.2 | Intron | NP_001107656.1 | ||||
NM_001301047.2 | Intron | NP_001287976.1 | ||||
NM_001301870.1 | Intron | NP_001288799.1 | ||||
NM_001301871.1 | Intron | NP_001288800.1 | ||||
NM_001301872.1 | Intron | NP_001288801.1 | ||||
NM_019041.6 | Intron | NP_061914.3 | ||||
XM_017010959.1 | Intron | XP_016866448.1 | ||||
XM_017010960.1 | Intron | XP_016866449.1 |