Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615365 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK8 - adenylate kinase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317958.1 | 189 | UTR 5 | NP_001304887.1 | |||
NM_001317959.1 | 189 | UTR 5 | NP_001304888.1 | |||
NM_152572.2 | 189 | UTR 5 | NP_689785.1 | |||
XM_005272169.2 | 189 | Intron | XP_005272226.1 | |||
XM_006716965.2 | 189 | Intron | XP_006717028.1 | |||
XM_011518277.2 | 189 | Intron | XP_011516579.1 | |||
XM_011518278.2 | 189 | Intron | XP_011516580.1 | |||
XM_017014308.1 | 189 | Intron | XP_016869797.1 |
SPACA9 - sperm acrosome associated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316897.1 | 189 | Intron | NP_001303826.1 | |||
NM_001316898.1 | 189 | Intron | NP_001303827.1 | |||
NM_001316900.1 | 189 | Intron | NP_001303829.1 | |||
NM_018956.4 | 189 | Intron | NP_061829.3 | |||
XM_017014231.1 | 189 | Intron | XP_016869720.1 |
Set Membership: |
HapMap |