Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATTTAATTTATTCTTTTGTGAGG[A/C]TATGTGGAGAACTAGCCAAGACCCG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300131 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLS3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLS3 - plastin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136025.4 | Intron | NP_001129497.1 | ||||
NM_001172335.2 | Intron | NP_001165806.1 | ||||
NM_001282337.1 | Intron | NP_001269266.1 | ||||
NM_001282338.1 | Intron | NP_001269267.1 | ||||
NM_005032.6 | Intron | NP_005023.2 | ||||
XM_011537534.1 | Intron | XP_011535836.1 |
PLS3-AS1 - PLS3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |