Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGGCCAAAACAATTATGACCCA[A/G]AGAAAAAACTTACAGTAGCTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600930 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPAM1 - sperm adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174044.1 | Intron | NP_001167515.1 | ||||
NM_001174045.1 | Intron | NP_001167516.1 | ||||
NM_001174046.1 | Intron | NP_001167517.1 | ||||
NM_003117.4 | Intron | NP_003108.2 | ||||
NM_153189.2 | Intron | NP_694859.1 |
Set Membership: |
HapMap |