Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCCCAGGGAGGCTCCTCGGGC[C/T]GGGGCACAGCTCTCAAGGGTGGCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164350 MIM: 603351 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OAS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
OAS1 - 2'-5'-oligoadenylate synthetase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OAS3 - 2'-5'-oligoadenylate synthetase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006187.3 | 212 | Missense Mutation | CGG,TGG | R,W 65 | NP_006178.2 | |
XM_005253889.3 | 212 | Missense Mutation | CGG,TGG | R,W 65 | XP_005253946.1 | |
XM_017019363.1 | 212 | Missense Mutation | CGG,TGG | R,W 65 | XP_016874852.1 |
Set Membership: |
HapMap |