Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTTACAGATTCATTATTAAATG[C/G]GCCATCTTTCAAGACAAAATAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603015 | ||||||||||||||||||||||||||||||||
Literature Links: |
MIR3609 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MIR3609 - microRNA 3609 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA28 - small Cajal body-specific RNA 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRRAP - transformation/transcription domain associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244580.1 | Intron | NP_001231509.1 | ||||
NM_003496.3 | Intron | NP_003487.1 |
Set Membership: |
HapMap |