Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615178 MIM: 191321 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KXD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
KXD1 - KxDL motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBA52 - ubiquitin A-52 residue ribosomal protein fusion product 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033930.2 | 1544 | UTR 3 | NP_001029102.1 | |||
NM_001321017.1 | 1544 | UTR 3 | NP_001307946.1 | |||
NM_001321018.1 | 1544 | UTR 3 | NP_001307947.1 | |||
NM_001321019.1 | 1544 | UTR 3 | NP_001307948.1 | |||
NM_001321020.1 | 1544 | UTR 3 | NP_001307949.1 | |||
NM_001321021.1 | 1544 | UTR 3 | NP_001307950.1 | |||
NM_001321022.1 | 1544 | UTR 3 | NP_001307951.1 | |||
NM_003333.4 | 1544 | UTR 3 | NP_003324.1 | |||
XM_005260052.2 | 1544 | Intron | XP_005260109.1 | |||
XM_005260053.3 | 1544 | Intron | XP_005260110.1 | |||
XM_005260054.2 | 1544 | Intron | XP_005260111.1 | |||
XM_006722871.2 | 1544 | UTR 3 | XP_006722934.1 | |||
XM_017027198.1 | 1544 | UTR 3 | XP_016882687.1 |