Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCGTTTTAGAGAGAAGGAAATT[A/G]AGAGACTGAAGTGGAGGAATTTACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605796 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TDRD1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TDRD1 - tudor domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VWA2 - von Willebrand factor A domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272046.1 | Intron | NP_001258975.1 | ||||
NM_001320804.1 | Intron | NP_001307733.1 | ||||
XM_011539754.2 | Intron | XP_011538056.1 | ||||
XM_011539757.2 | Intron | XP_011538059.1 | ||||
XM_017016177.1 | Intron | XP_016871666.1 | ||||
XM_017016178.1 | Intron | XP_016871667.1 | ||||
XM_017016179.1 | Intron | XP_016871668.1 | ||||
XM_017016180.1 | Intron | XP_016871669.1 | ||||
XM_017016181.1 | Intron | XP_016871670.1 |
Set Membership: |
HapMap |