Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTTACAGGTGTAAGCCACTATG[C/T]CTAGCCTGAGTACTATTATTATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602968 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCAS1 - breast carcinoma amplified sequence 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316361.2 | Intron | NP_001303290.1 | ||||
NM_001323347.1 | Intron | NP_001310276.1 | ||||
NM_003657.3 | Intron | NP_003648.2 | ||||
XM_005260587.2 | Intron | XP_005260644.1 | ||||
XM_005260589.2 | Intron | XP_005260646.1 | ||||
XM_005260590.2 | Intron | XP_005260647.1 | ||||
XM_005260591.2 | Intron | XP_005260648.1 | ||||
XM_005260593.2 | Intron | XP_005260650.1 | ||||
XM_005260594.2 | Intron | XP_005260651.1 | ||||
XM_005260595.2 | Intron | XP_005260652.1 | ||||
XM_011529090.2 | Intron | XP_011527392.1 | ||||
XM_011529091.2 | Intron | XP_011527393.1 | ||||
XM_017028110.1 | Intron | XP_016883599.1 | ||||
XM_017028111.1 | Intron | XP_016883600.1 | ||||
XM_017028112.1 | Intron | XP_016883601.1 |