Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGGGGCCCGTCCGCCATTGCCA[C/G]TGGCTCCCGCCCTTACTGGCAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605213 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDPK1 - 3-phosphoinositide dependent protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261816.1 | Intron | NP_001248745.1 | ||||
NM_002613.4 | Intron | NP_002604.1 | ||||
NM_031268.5 | Intron | NP_112558.2 | ||||
XM_011522521.2 | Intron | XP_011520823.1 | ||||
XM_011522523.2 | Intron | XP_011520825.1 | ||||
XM_017023271.1 | Intron | XP_016878760.1 | ||||
XM_017023272.1 | Intron | XP_016878761.1 | ||||
XM_017023273.1 | Intron | XP_016878762.1 | ||||
XM_017023274.1 | Intron | XP_016878763.1 | ||||
XM_017023275.1 | Intron | XP_016878764.1 | ||||
XM_017023276.1 | Intron | XP_016878765.1 | ||||
XM_017023277.1 | Intron | XP_016878766.1 |
Set Membership: |
HapMap |