Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602238 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EXOSC2 - exosome component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282708.1 | Intron | NP_001269637.1 | ||||
NM_001282709.1 | Intron | NP_001269638.1 | ||||
NM_014285.6 | Intron | NP_055100.2 | ||||
XM_005272176.2 | Intron | XP_005272233.1 | ||||
XM_006717023.2 | Intron | XP_006717086.1 | ||||
XM_017014558.1 | Intron | XP_016870047.1 |
Set Membership: |
HapMap |