Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGAACTAAAATCCAAGATTTTAT[A/G]GATTTCTCCAAATTGTGGCAATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX29 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SNX29 - sorting nexin 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032167.4 | Intron | NP_115543.3 | ||||
XM_005255682.3 | Intron | XP_005255739.1 | ||||
XM_005255683.3 | Intron | XP_005255740.1 | ||||
XM_011522738.2 | Intron | XP_011521040.1 | ||||
XM_011522741.2 | Intron | XP_011521043.1 | ||||
XM_011522743.2 | Intron | XP_011521045.1 | ||||
XM_011522744.2 | Intron | XP_011521046.1 | ||||
XM_017023871.1 | Intron | XP_016879360.1 | ||||
XM_017023872.1 | Intron | XP_016879361.1 | ||||
XM_017023873.1 | Intron | XP_016879362.1 | ||||
XM_017023874.1 | Intron | XP_016879363.1 | ||||
XM_017023875.1 | Intron | XP_016879364.1 | ||||
XM_017023876.1 | Intron | XP_016879365.1 | ||||
XM_017023877.1 | Intron | XP_016879366.1 | ||||
XM_017023878.1 | Intron | XP_016879367.1 |
Set Membership: |
HapMap |