Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAAGTGTCCCAGAGAAAGTAAA[T/G]TCTAAACTGAGAAGTGGAAGTGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USP42 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
USP42 - ubiquitin specific peptidase 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032172.2 | Intron | NP_115548.1 | ||||
XM_005249883.4 | Intron | XP_005249940.1 | ||||
XM_006715790.1 | Intron | XP_006715853.1 | ||||
XM_006715791.2 | Intron | XP_006715854.1 | ||||
XM_011515573.1 | Intron | XP_011513875.1 | ||||
XM_011515574.1 | Intron | XP_011513876.1 | ||||
XM_011515577.1 | Intron | XP_011513879.1 | ||||
XM_011515578.1 | Intron | XP_011513880.1 |
Set Membership: |
HapMap Validated |