Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCGGAGTTGGCGGTTCTGGCTG[A/T]TGGCCTGCTTCACCTGGAATCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605326 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAX1BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
TAX1BP1 - Tax1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079864.2 | Intron | NP_001073333.1 | ||||
NM_001206901.1 | Intron | NP_001193830.1 | ||||
NM_001206902.1 | Intron | NP_001193831.1 | ||||
NM_006024.6 | Intron | NP_006015.4 | ||||
XM_005249900.2 | Intron | XP_005249957.1 | ||||
XM_017012754.1 | Intron | XP_016868243.1 |