Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CECR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CECR5 - cat eye syndrome chromosome region, candidate 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017829.5 | Intron | NP_060299.4 | ||||
NM_033070.2 | Intron | NP_149061.1 | ||||
XM_005261236.3 | Intron | XP_005261293.1 | ||||
XM_011546126.2 | Intron | XP_011544428.1 | ||||
XM_011546127.1 | Intron | XP_011544429.1 | ||||
XM_017028781.1 | Intron | XP_016884270.1 | ||||
XM_017028782.1 | Intron | XP_016884271.1 | ||||
XM_017028783.1 | Intron | XP_016884272.1 | ||||
XM_017028784.1 | Intron | XP_016884273.1 |
CECR5-AS1 - CECR5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |