Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCGAGATTTTAAGAATTAAATG[A/G]GATAATGTATGTAAAGCACTTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616785 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHTF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PHTF2 - putative homeodomain transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127357.1 | Intron | NP_001120829.1 | ||||
NM_001127358.1 | Intron | NP_001120830.1 | ||||
NM_001127359.1 | Intron | NP_001120831.1 | ||||
NM_001127360.1 | Intron | NP_001120832.1 | ||||
NM_020432.4 | Intron | NP_065165.3 | ||||
XM_005250508.4 | Intron | XP_005250565.1 | ||||
XM_005250509.4 | Intron | XP_005250566.1 | ||||
XM_006716063.3 | Intron | XP_006716126.1 | ||||
XM_011516422.2 | Intron | XP_011514724.1 | ||||
XM_011516423.2 | Intron | XP_011514725.1 | ||||
XM_011516424.2 | Intron | XP_011514726.1 | ||||
XM_017012458.1 | Intron | XP_016867947.1 | ||||
XM_017012459.1 | Intron | XP_016867948.1 |
TMEM60 - transmembrane protein 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |