Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGGATTTCTATAGGAATCCCA[G/T]GAGGGTCTTACTGGAGGGTTGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608171 | ||||||||||||||||||||
Literature Links: |
CACNA2D4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNA2D4 - calcium voltage-gated channel auxiliary subunit alpha2delta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172364.4 | 484 | Intron | NP_758952.4 | |||
XM_011521041.2 | 484 | Intron | XP_011519343.1 |
LRTM2 - leucine rich repeats and transmembrane domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039029.2 | 484 | UTR 5 | NP_001034118.1 | |||
NM_001163925.1 | 484 | UTR 5 | NP_001157397.1 | |||
NM_001163926.1 | 484 | UTR 5 | NP_001157398.1 | |||
XM_011521015.1 | 484 | UTR 5 | XP_011519317.1 | |||
XM_017019848.1 | 484 | Intron | XP_016875337.1 |