Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614178 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRACR2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CRACR2A - calcium release activated channel regulator 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144958.1 | 2464 | Intron | NP_001138430.1 | |||
NM_032680.3 | 2464 | Intron | NP_116069.1 | |||
XM_006719021.3 | 2464 | UTR 3 | XP_006719084.1 | |||
XM_011521034.2 | 2464 | UTR 3 | XP_011519336.1 | |||
XM_011521036.2 | 2464 | UTR 3 | XP_011519338.1 | |||
XM_011521037.2 | 2464 | UTR 3 | XP_011519339.1 | |||
XM_011521038.2 | 2464 | UTR 3 | XP_011519340.1 |
Set Membership: |
HapMap Validated |