Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCGCCACCTGCCTCCCGGCCTG[C/T]TCCTTTTGTTCCCCCTCCCACTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITFG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITFG2 - integrin alpha FG-GAP repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018463.3 | 1338 | Intron | NP_060933.3 | |||
XM_005253707.4 | 1338 | Intron | XP_005253764.1 | |||
XM_005253709.4 | 1338 | Intron | XP_005253766.1 | |||
XM_017019641.1 | 1338 | Intron | XP_016875130.1 |
LOC100507424 - uncharacterized LOC100507424 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRIP2 - nuclear receptor interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031474.2 | 1338 | Intron | NP_113662.1 | |||
XM_005253797.3 | 1338 | Intron | XP_005253854.1 | |||
XM_005253798.3 | 1338 | Intron | XP_005253855.1 | |||
XM_011521031.2 | 1338 | Intron | XP_011519333.1 | |||
XM_017020000.1 | 1338 | UTR 3 | XP_016875489.1 | |||
XM_017020001.1 | 1338 | Intron | XP_016875490.1 |