Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGCCATCGTGCCCAGCCACTCT[C/T]ACTTTAATACTATGAAAAATGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf75 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C17orf75 - chromosome 17 open reading frame 75 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022344.3 | 2894 | UTR 3 | NP_071739.2 | |||
XM_005258022.3 | 2894 | UTR 3 | XP_005258079.1 | |||
XM_017024940.1 | 2894 | UTR 3 | XP_016880429.1 | |||
XM_017024941.1 | 2894 | UTR 3 | XP_016880430.1 |
RHBDL3 - rhomboid like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |