Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf75 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf75 - chromosome 17 open reading frame 75 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR632 - microRNA 632 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF207 - zinc finger protein 207 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032293.2 | Intron | NP_001027464.1 | ||||
NM_001098507.1 | Intron | NP_001091977.1 | ||||
NM_003457.3 | Intron | NP_003448.1 | ||||
XM_011525185.2 | Intron | XP_011523487.1 | ||||
XM_017025019.1 | Intron | XP_016880508.1 | ||||
XM_017025020.1 | Intron | XP_016880509.1 |