Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102981 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADCYAP1R1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ADCYAP1R1 - ADCYAP receptor type I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001118.4 | Intron | NP_001109.2 | ||||
NM_001199635.1 | Intron | NP_001186564.1 | ||||
NM_001199636.1 | Intron | NP_001186565.1 | ||||
NM_001199637.1 | Intron | NP_001186566.1 | ||||
XM_005249618.4 | Intron | XP_005249675.1 | ||||
XM_006715645.2 | Intron | XP_006715708.1 | ||||
XM_017011736.1 | Intron | XP_016867225.1 | ||||
XM_017011737.1 | Intron | XP_016867226.1 | ||||
XM_017011738.1 | Intron | XP_016867227.1 |
Set Membership: |
HapMap Validated |