Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612013 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CC2D2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CC2D2A - coiled-coil and C2 domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080522.2 | Intron | NP_001073991.2 | ||||
NM_001164720.1 | Intron | NP_001158192.1 | ||||
NM_020785.2 | Intron | NP_065836.2 | ||||
XM_005248177.1 | Intron | XP_005248234.1 | ||||
XM_011513872.2 | Intron | XP_011512174.1 | ||||
XM_011513874.2 | Intron | XP_011512176.1 | ||||
XM_017008482.1 | Intron | XP_016863971.1 |