Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATTAACTGTGAGACGAAGTGAG[A/G]CCAAACAGTCTGGGAATTATGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929770 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929770 - uncharacterized LOC101929770 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SUPT3H - SPT3 homolog, SAGA and STAGA complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261823.1 | Intron | NP_001248752.1 | ||||
NM_003599.3 | Intron | NP_003590.1 | ||||
NM_181356.2 | Intron | NP_852001.1 | ||||
XM_011514949.2 | Intron | XP_011513251.1 | ||||
XM_011514950.1 | Intron | XP_011513252.1 | ||||
XM_011514951.2 | Intron | XP_011513253.1 | ||||
XM_011514952.2 | Intron | XP_011513254.1 | ||||
XM_011514953.2 | Intron | XP_011513255.1 | ||||
XM_011514954.2 | Intron | XP_011513256.1 | ||||
XM_017011369.1 | Intron | XP_016866858.1 | ||||
XM_017011370.1 | Intron | XP_016866859.1 | ||||
XM_017011371.1 | Intron | XP_016866860.1 | ||||
XM_017011372.1 | Intron | XP_016866861.1 | ||||
XM_017011373.1 | Intron | XP_016866862.1 | ||||
XM_017011374.1 | Intron | XP_016866863.1 |